Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Mettler Toledo Economic & Easy pH electrode
Synonym(s):
Mettler-Toledo Economic & Easy pH electrode
Products
Manufacturer
Diameter (mm)
Length (mm)
Membrane Material
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
MT51340242
Mettler Toledo Economic & Easy pH electrode,
BNC/RCA connection, glass, U membrane, plastic, POM shaft, integrated temperature sensor
Expand
View Pricing
MT30089747
Mettler Toledo Economic & Easy pH electrode,
BNC connection, ceramic membrane, glass shaft
Expand
View Pricing
MT51340330
Mettler Toledo Economic & Easy pH electrode,
BNC connection, glass, U membrane, plastic, POM shaft, Two nylon diaphragms
Expand
View Pricing
MT51340331
Mettler Toledo Economic & Easy pH electrode,
BNC connection, glass, HA membrane, glass shaft
Expand
View Pricing
MT51340332
Mettler Toledo Economic & Easy pH electrode,
BNC connection, glass, HA membrane, glass shaft, Suitable for TRIS buffers
Expand
View Pricing