Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Methylmagnesium bromide solution
Linear Formula:
CH
3
MgBr
CAS Number:
75-16-1
Molecular Weight:
119.24
Products
Product Category
organic bases (4)
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Quality Segment
Reaction Type
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
189898
Methylmagnesium bromide solution,
3.0 M in diethyl ether
Expand
View Pricing
703583
Methylmagnesium bromide solution,
~3.4 M in 2-methyltetrahydrofuran
Expand
View Pricing
282235
Methylmagnesium bromide solution,
1.4 M in THF: toluene (1:3)
Expand
View Pricing
257087
Methylmagnesium bromide solution,
3.0 M in diethyl ether
Expand
View Pricing
302430
Methylmagnesium bromide solution,
1.0 M in dibutyl ether
Expand
View Pricing