
Methylenediphosphonic acid

Synonym(s):
MDP, Medronic acid, Methylenediphosphonic acid
Linear Formula:
CH2[P(O)(OH)2]2
CAS Number:
Molecular Weight:
176.00
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-3 of 3