
Methylcobalamin

Synonym(s):
Methylcobalamin
Empirical Formula (Hill Notation):
C63H91CoN13O14P
CAS Number:
Molecular Weight:
1344.38
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3