
Methazolamide

Synonym(s):
5-Acetylimino-4-methyl-·2-1,3,4-thiadiazoline-2-sulfonamide, N-(4-Methyl-2-sulfamoyl-Δ2-1,3,4-thiadiazolin-5-ylidene) acetamide, L 584601, N-[5-(Aminosulfonyl)-3-methyl-1,3,4-thiadiazol-2(3H)-ylidene]acetamide, Methazolamide
Empirical Formula (Hill Notation):
C5H8N4O3S2
CAS Number:
Molecular Weight:
236.27
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-3 of 3