Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Metanil Yellow
Synonym(s):
3-(4-Anilinophenylazo)benzenesulfonic acid sodium salt, Acid Yellow 36, Metanil yellow
Empirical Formula (Hill Notation):
C
18
H
14
N
3
NaO
3
S
CAS Number:
587-98-4
Molecular Weight:
375.38
EC Number:
209-608-2
Products
Brand
Sigma-Aldrich (1)
Supelco (2)
Color
Formula Weight
pH Value
Physical Form
Purity
Agency
Format
Chemical Composition
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
202029
Metanil Yellow,
Dye content 70 %
Expand
View Pricing
1.59267
Metanil Yellow,
pH indicator, Reag. Ph Eur
Expand
View Pricing
44426
Metanil Yellow,
analytical standard
Expand
View Pricing