Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Maize GMO Standard ERM-BF423
Synonym(s):
GVO Standard ERM-BF423, Maize MIR604
Products
Manufacturer
Format
Chemical Composition
Application
Reference Material Type
Special Grade
certified reference material (4)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
ERMBF423C
Maize GMO Standard ERM-BF423,
1% MIR604, for the detection of GMO maize (MIR604) with PCR
Expand
View Pricing
ERMBF423B
Maize GMO Standard ERM-BF423,
0.1% MIR604, for the detection of GMO maize (MIR604) with PCR
Expand
View Pricing
ERMBF423D
Maize GMO Standard ERM-BF423,
10% MIR604, for the detection of GMO maize (MIR604) with PCR
Expand
View Pricing
ERMBF423A
Maize GMO Standard ERM-BF423,
0% MIR604, for the detection of GMO maize (MIR604) with PCR
Expand
View Pricing