Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Magic Touch 2
™
ice bucket with lid
Products
Manufacturer
Capacity (L)
Color
Package Size
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
BAM168072002
Magic Touch 2
™
ice bucket with lid,
capacity 2.5 L, black, pack of 1 ea
Expand
View Pricing
BAM168072003
Magic Touch 2
™
ice bucket with lid,
capacity 2.5 L, red, pack of 1 ea
Expand
View Pricing
BAM168072001
Magic Touch 2
™
ice bucket with lid,
capacity 2.5 L, blue, pack of 1 ea
Expand
View Pricing
BAM168074002
Magic Touch 2
™
ice bucket with lid,
capacity 4.0 L, black, pack of 1 ea
Expand
View Pricing
BAM168074001
Magic Touch 2
™
ice bucket with lid,
capacity 4.0 L, blue, pack of 1 ea
Expand
View Pricing
BAM168074003
Magic Touch 2
™
ice bucket with lid,
capacity 4.0 L, red, pack of 1 ea
Expand
View Pricing
BAM168072004
Magic Touch 2
™
ice bucket with lid,
capacity 2.5 L, green, pack of 1 ea
Expand
View Pricing
BAM168074004
Magic Touch 2
™
ice bucket with lid,
capacity 4.0 L, green, pack of 1 ea
Expand
View Pricing