
Loperamide Related Compound F

Synonym(s):
Loperamide N-oxide, trans-4-(4-Chlorophenyl)-4-hydroxy-N,N-dimethyl-α,α-diphenyl-1-piperidinebutanamide 1-oxide
Empirical Formula (Hill Notation):
C29H33ClN2O3
CAS Number:
Molecular Weight:
493.04
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2