Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
L-Norleucine
Synonym(s):
(
S
)-(+)-2-Aminohexanoic acid, (
S
)-2-Aminocaproic acid
Linear Formula:
CH
3
(CH
2
)
3
CH(NH
2
)CO
2
H
CAS Number:
327-57-1
Molecular Weight:
131.17
EC Number:
206-321-4
Products
Product Category
amino acids (2)
Brand
Sigma-Aldrich (1)
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Optical Isomer
Amino Acid
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
N6877
L-Norleucine,
≥98% (TLC)
Expand
View Pricing
N8513
L-Norleucine,
suitable for amino acid analysis, BioReagent
Expand
View Pricing