
L-Menthyl lactate

Empirical Formula (Hill Notation):
C13H24O3
CAS Number:
Molecular Weight:
228.33
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Agency
Quality Segment
Application
Optical Isomer
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing