![L-Leucinamide hydrochloride 99%](/deepweb/assets/sigmaaldrich/product/structures/224/498/0b731a78-1943-43e0-8294-77b8c4424cd5/640/0b731a78-1943-43e0-8294-77b8c4424cd5.png)
L-Leucinamide hydrochloride
![L-Leucinamide hydrochloride 99%](/deepweb/assets/sigmaaldrich/product/structures/224/498/0b731a78-1943-43e0-8294-77b8c4424cd5/640/0b731a78-1943-43e0-8294-77b8c4424cd5.png)
Linear Formula:
(CH3)2CHCH2CH(NH2)CONH2·HCl
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing