Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lithium 3,5-diiodosalicylate
Synonym(s):
2-Hydroxy-3,5-diiodobenzoic acid lithium salt, 3,5-Diiodo-2-hydroxybenzoic acid lithium salt, 3,5-Diiodosalicylic acid lithium salt, LIS
Empirical Formula (Hill Notation):
C
7
H
3
I
2
LiO
3
CAS Number:
653-14-5
Molecular Weight:
395.85
EC Number:
211-496-5
Products
Product Category
detergents (1)
Brand
Supelco (1)
Formula Weight
Format
Quality Segment
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
D3635
Lithium 3,5-diiodosalicylate,
analytical standard
Expand
View Pricing