Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lipopolysaccharides from
Escherichia coli
O26:B6
Synonym(s):
LPS
EC Number:
297-473-0
Products
Product Category
carbohydrates (5)
lipopolysaccarides (4)
Brand
Sigma-Aldrich (5)
Biological Source
Color
Physical Form
Product Line
Quality Segment
Sterilization Method
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
L2654
Lipopolysaccharides from
Escherichia coli
O26:B6,
γ-irradiated, BioXtra, suitable for cell culture
Expand
View Pricing
L8274
Lipopolysaccharides from
Escherichia coli
O26:B6,
≥10,000 EU/mg, purified by phenol extraction
Expand
View Pricing
L5543
Lipopolysaccharides from
Escherichia coli
O26:B6,
Ready Made solution, 1 mg/mL, 0.2 μm filtered
Expand
View Pricing
L2762
Lipopolysaccharides from
Escherichia coli
O26:B6,
purified by gel-filtration chromatography
Expand
View Pricing
L3755
Lipopolysaccharides from
Escherichia coli
O26:B6,
purified by trichloroacetic acid extraction
Expand
View Pricing