Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Linear polyethylenimine-
block
-poly(ethylene glycol)
Linear Formula:
CH
3
(C
2
H
4
O)
n
(C
2
H
5
N)
m
OH
Products
Brand
Sigma-Aldrich (8)
Color
Physical Form
Quality Segment
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
933910
Linear polyethylenimine-
block
-poly(ethylene glycol),
PEG average M
n
5,000, PEI average M
n
30,000
Expand
View Pricing
933724
Linear polyethylenimine-
block
-poly(ethylene glycol),
PEG average M
n
5,000, PEI average M
n
10,000
Expand
View Pricing
927309
Linear polyethylenimine-
block
-poly(ethylene glycol),
PEG average M
n
2,000, PEI average M
n
20,000
Expand
View Pricing
929867
Linear polyethylenimine-
block
-poly(ethylene glycol),
PEG average M
n
2,000, PEI average M
n
30,000
Expand
View Pricing
933732
Linear polyethylenimine-
block
-poly(ethylene glycol),
PEG average M
n
5,000, PEI average M
n
5,000
Expand
View Pricing
927317
Linear polyethylenimine-
block
-poly(ethylene glycol),
PEG average M
n
750, PEI average M
n
15,000
Expand
View Pricing
927287
Linear polyethylenimine-
block
-poly(ethylene glycol),
PEG average M
n
750, PEI average M
n
10,000
Expand
View Pricing
927295
Linear polyethylenimine-
block
-poly(ethylene glycol),
PEG average M
n
2,000, PEI average M
n
10,000
Expand
View Pricing