Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
LiChrosorb
®
Si 100 (10 µm) HPLC Columns
Synonym(s):
Silica gel
CAS Number:
112926-00-8
Products
Brand
Supelco (1)
Boiling Point (°C)
Melting Point (°C)
pH Value
Product Line
Agency
USP Suitability
Separation Technique
Column Material
Particle Size (µm)
Pore Size (Å)
Technique
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
1.09309
LiChrosorb
®
Si 100 (10 µm) HPLC Columns,
HPLC sorbent, CAS 7631-86-9, EC Number 231-545-4, chemical formula SiO₂.
Expand
View Pricing