
L-Glutathione oxidized disodium salt

Synonym(s):
GSSG
Empirical Formula (Hill Notation):
C20H30N6Na2O12S2
CAS Number:
Molecular Weight:
656.59
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Optical Isomer
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing