Skip to Content
Merck
L-Cysteic acid monohydrate ≥99.0% (T)

L-Cysteic acid monohydrate

L-Cysteic acid monohydrate ≥99.0% (T)
Synonym(s):
(R)-2-Amino-3-sulfopropionic acid
Linear Formula:
HO3SCH2CH(NH2)CO2H·H2O
CAS Number:
Molecular Weight:
187.17

Products

Product Category
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Optical Isomer
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing