Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
L-Carnitine inner salt
Synonym(s):
(−)-(
R
)-3-Hydroxy-4-(trimethylammonio)butyrate, Vitamin B
T
Linear Formula:
(CH
3
)
3
N
+
CH
2
CH(OH)CH
2
COO
-
CAS Number:
541-15-1
Molecular Weight:
161.20
EC Number:
208-768-0
Products
Product Category
amino acids (1)
Brand
Sigma-Aldrich (1)
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Optical Isomer
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
C0158
L-Carnitine inner salt,
≥98%
Expand
View Pricing