
Lactose monohydrate

Synonym(s):
D-Lactose monohydrate, β-D-Gal-(1→4)-D-Glc, 4-O-β-D-Galactopyranosyl-D-glucose, Milk sugar
Empirical Formula (Hill Notation):
C12H22O11 · H2O
CAS Number:
Molecular Weight:
360.31
Products
Product Category
Brand
Formula Weight
Manufacturer
Agency
Format
Analyte Suitability
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3