Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lactic Dehydrogenase, recombinant from
E. coli
Synonym(s):
(
S
)-Lactate: NAD
+
oxidoreductase, L-Lactate Dehydrogenase, Lactate
CAS Number:
9001-60-9
EC Number:
232-617-8
Products
Product Category
enzymes (1)
Brand
Sigma-Aldrich (1)
Physical Form
Recombinant Host
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
59747
Lactic Dehydrogenase, recombinant from
E. coli
,
≥90 U/mg
Expand
View Pricing