Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Kontes
®
NMR tubes 5 mm diam., precision
Products
Product Category
tubes (23)
Diameter (mm)
Length (in)
Color
Frequency (MHz)
Filter Products
Showing 1-20 of 23
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
KN8972350008
Kontes
®
NMR tubes 5 mm diam., precision,
frequency 400 MHz, L × camber × concentricity 8 in. × 15 μm × 25 μm
Expand
View Pricing
KN8972350000
Kontes
®
NMR tubes 5 mm diam., precision,
frequency 400 MHz, L × camber × concentricity 7 in. × 13 μm × 25 μm
Expand
View Pricing
KN8972400000
Kontes
®
NMR tubes 5 mm diam., precision,
frequency 500 MHz, L × camber × concentricity 7 in. × 6 μm × 13 μm
Expand
View Pricing
KN8972050009
Kontes
®
NMR tubes 5 mm diam., precision,
frequency 100 MHz, L × camber × concentricity 9 in. × 51 μm × 64 μm
Expand
View Pricing
KN8972410008
Kontes
®
NMR tubes 5 mm diam., precision,
frequency 600-700 MHz, L × camber × concentricity 8 in. × 9 μm × 13 μm
Expand
View Pricing
KN8972250008
Kontes
®
NMR tubes 5 mm diam., precision,
frequency 200 MHz, L × camber × concentricity 8 in. × 15 μm × 51 μm
Expand
View Pricing
KN8972000009
Kontes
®
NMR tubes 5 mm diam., precision,
frequency 100 MHz, L × camber × concentricity 9 in. × 51 μm × 76 μm
Expand
View Pricing
KN8972250009
Kontes
®
NMR tubes 5 mm diam., precision,
frequency 200 MHz, L × camber × concentricity 9 in. × 18 μm × 51 μm
Expand
View Pricing
KN8972000000
Kontes
®
NMR tubes 5 mm diam., precision,
frequency 100 MHz, L × camber × concentricity 7 in. × 51 μm × 76 μm
Expand
View Pricing
KN8972000008
Kontes
®
NMR tubes 5 mm diam., precision,
frequency 100 MHz, L × camber × concentricity 8 in. × 51 μm × 76 μm
Expand
View Pricing
KN8972050008
Kontes
®
NMR tubes 5 mm diam., precision,
frequency 100 MHz, L × camber × concentricity 8 in. × 51 μm × 64 μm
Expand
View Pricing
KN8972503000
Kontes
®
NMR tubes 5 mm diam., precision,
frequency 900 MHz, L × camber × concentricity 7 in. × 3.8 μm × 5 μm
Expand
View Pricing
KN8972300008
Kontes
®
NMR tubes 5 mm diam., precision,
frequency 300 MHz, L × camber × concentricity 8 in. × 28 μm × 38 μm
Expand
View Pricing
KN8972453000
Kontes
®
NMR tubes 5 mm diam., precision,
frequency 800 MHz, L × camber × concentricity 7 in. × 3.8 μm × 5 μm
Expand
View Pricing
KN8972410000
Kontes
®
NMR tubes 5 mm diam., precision,
frequency 600-700 MHz, L × camber × concentricity 7 in. × 6 μm × 13 μm
Expand
View Pricing
KN8972400009
Kontes
®
NMR tubes 5 mm diam., precision,
frequency 500 MHz, L × camber × concentricity 9 in. × 11 μm × 13 μm
Expand
View Pricing
KN8972453008
Kontes
®
NMR tubes 5 mm diam., precision,
frequency 800 MHz, L × camber × concentricity 8 in. × 3.8 μm × 5 μm
Expand
View Pricing
KN8972400008
Kontes
®
NMR tubes 5 mm diam., precision,
frequency 500 MHz, L × camber × concentricity 8 in. × 9 μm × 13 μm
Expand
View Pricing
KN8972350009
Kontes
®
NMR tubes 5 mm diam., precision,
frequency 400 MHz, L × camber × concentricity 9 in. × 18 μm × 25 μm
Expand
View Pricing
KN8972300000
Kontes
®
NMR tubes 5 mm diam., precision,
frequency 300 MHz, L × camber × concentricity 7 in. × 25 μm × 38 μm
Expand
View Pricing
Page 1 of 2
Page 1 of 2