Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Kontes
®
NMR tubes 10 mm diam., disposable grade
Products
Product Category
tubes (2)
Diameter (mm)
Length (in)
Color
Frequency (MHz)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
KN8973000000
Kontes
®
NMR tubes 10 mm diam., disposable grade,
frequency 100 MHz, L 7 in.
Expand
View Pricing
KN8973000008
Kontes
®
NMR tubes 10 mm diam., disposable grade,
frequency 100 MHz, L 8 in.
Expand
View Pricing