Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
KIMBLE
®
KIM-KAP
®
push cap
Products
Product Category
caps (21)
Diameter (mm)
Color
Package Size
Sterility
Filter Products
Showing 1-20 of 21
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
DWK73660-16
KIMBLE
®
KIM-KAP
®
push cap,
assorted colors polypropylene cap, cap diam. (16 mm)
Expand
View Pricing
DWK73660-25
KIMBLE
®
KIM-KAP
®
push cap,
natural polypropylene cap, cap diam. (25 mm)
Expand
View Pricing
DWK73660-20
KIMBLE
®
KIM-KAP
®
push cap,
assorted colors polypropylene cap, cap diam. (20 mm)
Expand
View Pricing
DWK73665-16
KIMBLE
®
KIM-KAP
®
push cap,
blue polypropylene cap, cap diam. (16 mm)
Expand
View Pricing
DWK73665-20
KIMBLE
®
KIM-KAP
®
push cap,
blue polypropylene cap, cap diam. (20 mm)
Expand
View Pricing
DWK73660-13
KIMBLE
®
KIM-KAP
®
push cap,
assorted colors polypropylene cap, cap diam. (13 mm)
Expand
View Pricing
DWK73660-18
KIMBLE
®
KIM-KAP
®
push cap,
assorted colors polypropylene cap, cap diam. (18 mm)
Expand
View Pricing
DWK73663-16
KIMBLE
®
KIM-KAP
®
push cap,
green polypropylene cap, cap diam. (16 mm)
Expand
View Pricing
DWK73665-18
KIMBLE
®
KIM-KAP
®
push cap,
blue polypropylene cap, cap diam. (18 mm)
Expand
View Pricing
DWK73660-38
KIMBLE
®
KIM-KAP
®
push cap,
assorted colors polypropylene cap, cap diam. (38 mm)
Expand
View Pricing
DWK73664-13
KIMBLE
®
KIM-KAP
®
push cap,
yellow polypropylene cap, cap diag. 13 mm
Expand
View Pricing
DWK73665-25
KIMBLE
®
KIM-KAP
®
push cap,
blue polypropylene cap, cap diam. (25 mm)
Expand
View Pricing
DWK73663-20
KIMBLE
®
KIM-KAP
®
push cap,
green polypropylene cap, cable diam. (20 mm)
Expand
View Pricing
DWK73662-20
KIMBLE
®
KIM-KAP
®
push cap,
red polypropylene cap, cap diam. (20 mm)
Expand
View Pricing
DWK73662-25
KIMBLE
®
KIM-KAP
®
push cap,
red polypropylene cap, cap diam. (25 mm)
Expand
View Pricing
DWK73663-25
KIMBLE
®
KIM-KAP
®
push cap,
green polypropylene cap, cap diam. (25 mm)
Expand
View Pricing
DWK73664-20
KIMBLE
®
KIM-KAP
®
push cap,
yellow polypropylene cap, cap diam. (20 mm)
Expand
View Pricing
DWK73662-13
KIMBLE
®
KIM-KAP
®
push cap,
red polypropylene cap, cap diam. (13 mm)
Expand
View Pricing
DWK73663-13
KIMBLE
®
KIM-KAP
®
push cap,
green polypropylene cap, cap diam. (13 mm)
Expand
View Pricing
DWK73662-16
KIMBLE
®
KIM-KAP
®
push cap,
red polypropylene cap, cap diam. (16 mm)
Expand
View Pricing
Page 1 of 2
Page 1 of 2