![Isophorone diisocyanate 98%, mixture of isomers](/deepweb/assets/sigmaaldrich/product/structures/635/798/b22afd5e-bf34-401e-bbc5-fe37f43744a8/640/b22afd5e-bf34-401e-bbc5-fe37f43744a8.png)
Isophorone diisocyanate
![Isophorone diisocyanate 98%, mixture of isomers](/deepweb/assets/sigmaaldrich/product/structures/635/798/b22afd5e-bf34-401e-bbc5-fe37f43744a8/640/b22afd5e-bf34-401e-bbc5-fe37f43744a8.png)
Synonym(s):
5-Isocyanato-1-(isocyanatomethyl)-1,3,3-trimethylcyclohexane
Linear Formula:
OCNC6H7(CH3)3CH2NCO
Molecular Weight:
222.28
EC Number:
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing