Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Iron/Silicon
Products
Manufacturer
Physical Form
Particle Size (µm)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
GF58129308
Iron/Silicon,
powder, Fe 91%/Si 9%, max. particle size 150 micron, weight 20 g, condition gas atomized
Expand
View Pricing
GF52403183
Iron/Silicon,
powder, Fe 91%/Si 9%, max. particle size 150 micron, weight 500 g, condition gas atomized
Expand
View Pricing
GF23624018
Iron/Silicon,
powder, Fe 91%/Si 9%, max. particle size 150 micron, weight 50 g, condition gas atomized
Expand
View Pricing
GF93876788
Iron/Silicon,
powder, Fe91%/Si 9%, 150 max. part. size (micron), weight 200 g, condition gas atomized
Expand
View Pricing
GF60529327
Iron/Silicon,
powder, 150 max. part. size (micron), Fe91%/Si 9%, weight 100 g, condition gas atomized
Expand
View Pricing