Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Inconel
®
alloy 600 - heat resisting alloy, Ni72Cr16Fe8
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Filter Products
Showing 1-20 of 201
Page 1 of 11
Page 1 of 11
Compare
Product Number
Description
Pricing
GF21058916
Inconel
®
alloy 600 - heat resisting alloy, Ni72Cr16Fe8,
rod, 1000mm x 6.35mm diameter, annealed
Expand
View Pricing
GF45461943
Inconel
®
alloy 600 - heat resisting alloy, Ni72Cr16Fe8,
rod, 500mm x 2.0mm diameter, annealed
Expand
View Pricing
GF46485995
Inconel
®
alloy 600 - heat resisting alloy, Ni72Cr16Fe8,
rod, 200mm x 2.0mm diameter, annealed
Expand
View Pricing
GF67601055
Inconel
®
alloy 600 - heat resisting alloy, Ni72Cr16Fe8,
rod, 200mm x 40mm diameter, annealed
Expand
View Pricing
GF19340658
Inconel
®
alloy 600 - heat resisting alloy, Ni72Cr16Fe8,
foil, 10mm disks, 0.0125mm thickness, annealed
Expand
View Pricing
GF40168470
Inconel
®
alloy 600 - heat resisting alloy, Ni72Cr16Fe8,
rod, 500mm x 6.35mm diameter, annealed
Expand
View Pricing
GF68054878
Inconel
®
alloy 600 - heat resisting alloy, Ni72Cr16Fe8,
20m wire, 0.25mm diameter, hard
Expand
View Pricing
GF22131021
Inconel
®
alloy 600 - heat resisting alloy, Ni72Cr16Fe8,
20m wire, 0.5mm diameter, hard
Expand
View Pricing
GF06129389
Inconel
®
alloy 600 - heat resisting alloy, Ni72Cr16Fe8,
foil, 15mm disks, 0.075mm thickness, annealed
Expand
View Pricing
GF99283348
Inconel
®
alloy 600 - heat resisting alloy, Ni72Cr16Fe8,
100mm tube, 12.7mm outside diameter, 1.63mm wall thickness, 9.44mm inside diameter, annealed
Expand
View Pricing
GF43776434
Inconel
®
alloy 600 - heat resisting alloy, Ni72Cr16Fe8,
100mm tube, 9.5mm outside diameter, 1.7mm wall thickness, 6.1mm inside diameter, annealed
Expand
View Pricing
GF23081420
Inconel
®
alloy 600 - heat resisting alloy, Ni72Cr16Fe8,
foil, 0.5m coil, 76mm coil width, 0.1mm thickness, annealed
Expand
View Pricing
GF72205343
Inconel
®
alloy 600 - heat resisting alloy, Ni72Cr16Fe8,
foil, 50 x 50mm, 0.1mm thickness, annealed
Expand
View Pricing
GF74013735
Inconel
®
alloy 600 - heat resisting alloy, Ni72Cr16Fe8,
foil, 150 x 150mm, 0.025mm thickness, annealed, light tight (lt)
Expand
View Pricing
GF36891759
Inconel
®
alloy 600 - heat resisting alloy, Ni72Cr16Fe8,
rod, 500mm x 3.2mm diameter, annealed
Expand
View Pricing
GF35751157
Inconel
®
alloy 600 - heat resisting alloy, Ni72Cr16Fe8,
20m wire, 0.60mm diameter, hard
Expand
View Pricing
GF44461021
Inconel
®
alloy 600 - heat resisting alloy, Ni72Cr16Fe8,
rod, 50mm x 40mm diameter, annealed
Expand
View Pricing
GF65363867
Inconel
®
alloy 600 - heat resisting alloy, Ni72Cr16Fe8,
rod, 1000mm x 3.2mm diameter, annealed
Expand
View Pricing
GF15769705
Inconel
®
alloy 600 - heat resisting alloy, Ni72Cr16Fe8,
200mm tube, 1.96mm outside diameter, 0.37mm wall thickness, 1.22mm inside diameter
Expand
View Pricing
GF34776591
Inconel
®
alloy 600 - heat resisting alloy, Ni72Cr16Fe8,
100m wire, 0.40mm diameter, hard
Expand
View Pricing
Page 1 of 11
Page 1 of 11