
Imperatorin

Synonym(s):
8-Isopentenyloxypsoralene, 9-(3-Methylbut-2-enyloxy)-7H-furo[3,2-g]chromen-7-one, 9-[(3-Methyl-2-buten-1-yl)oxy]-7h-furo[3,2-g][1]benzopyran-7-one, Ammidin, Marmelide, Marmelosin, NSC 402949
Empirical Formula (Hill Notation):
C16H14O4
CAS Number:
Molecular Weight:
270.28
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing