Skip to Content
Merck
Imazaquin PESTANAL®, analytical standard

Imazaquin

Imazaquin PESTANAL®, analytical standard
Synonym(s):
2-(4-Isopropyl-4-methyl-5-oxo-4,5-dihydro-1H-imidazol-2-yl)-quinoline-3-carboxylic acid
Empirical Formula (Hill Notation):
C17H17N3O3
CAS Number:
Molecular Weight:
311.34

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique