Skip to Content

End of year dates: Orders must be billed by 13/12. Merck Argentina will be closed from 23/12 to 02/01.

Merck
vibrant-m

Hyoscyamine sulfate

vibrant-m
Synonym(s):
S)-α-(Hydroxymethyl)benzeneacetic acid (3-endo)-8-methyl-8-azabicyclo[3.2.1]oct-3-yl ester sulfate hydrate (2:1:2)
Empirical Formula (Hill Notation):
C34H46N2O6 · H2SO4 · 2H2O
CAS Number:
Molecular Weight:
712.85

Products

Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing