Skip to Content
Merck
Hexylene glycol

Hexylene glycol

Hexylene glycol
Synonym(s):
(±)-2-Methyl-2,4-pentanediol, MPD, Hexylene glycol
Linear Formula:
CH3CH(OH)CH2C(CH3)2OH
CAS Number:
Molecular Weight:
118.17
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Manufacturer
pH Value
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reference Material Type
Special Grade