Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hettich
®
centrifuge adapters
Products
Product Category
adapters (19)
Manufacturer
Package Size
Technique
Filter Products
Showing 1-19 of 19
Compare
Product Number
Description
Pricing
Z653292
Hettich
®
centrifuge adapters,
for ROTOFIX 32A, ROTINA 38/38R and UNIVERSAL 320/320R, Adapter for 15 ml Falcon tubes (17x120 mm)
Expand
View Pricing
Z657905
Hettich
®
centrifuge adapters,
adapter for 50 ml tubes 34x100 mm
Expand
View Pricing
Z631159
Hettich
®
centrifuge adapters,
for 50 mL conical tubes (29x115 mm)
Expand
View Pricing
Z601586
Hettich
®
centrifuge adapters,
for ROTOFIX 32A and UNIVERSAL 320/320R Centrifuges, Adapter for 15 mL Falcon tubes (17x120 mm)
Expand
View Pricing
Z601594
Hettich
®
centrifuge adapters,
for ROTINA 38/38R and ROTINA 420/420R Centrifuges, Adapter for 50 mL Falcon tubes (29x115 mm)
Expand
View Pricing
Z657875
Hettich
®
centrifuge adapters,
for 8.5-10 mL blood/urine tubes (16x100 mm) and 15mL round bottom tubes (17x100 mm)
Expand
View Pricing
Z665592
Hettich
®
centrifuge adapters,
for ROTINA 38/38R, ROTINA 420/420R and ROTANTA 460/460R Centrifuges, Adapter for 50 ml Falcon tubes (29x115 mm)
Expand
View Pricing
Z657867
Hettich
®
centrifuge adapters,
for 4.5-5 mL blood/urine tubes (11x92 mm) and 7ml round bottom tubes (12x100 mm)
Expand
View Pricing
Z665606
Hettich
®
centrifuge adapters,
for ROTINA 38/38R, ROTINA 420/420R and ROTANTA 460/460R, Adapter for 1.5-2.0 mL tubes (11x 38 mm)
Expand
View Pricing
Z601578
Hettich
®
centrifuge adapters,
for ROTINA 38/38R and ROTINA 420/420R Centrifuges, Adapter for 9-15 mL tubes (14-17x100 mm)
Expand
View Pricing
Z653454
Hettich
®
centrifuge adapters,
for ROTOFIX 32A and UNIVERSAL 320/320R, Adapter (rubber insert) for 100 mL tubes (44x100 mm)
Expand
View Pricing
Z653403
Hettich
®
centrifuge adapters,
for 7.5-10 mL blood/urine tubes (15-16x92-102 mm) and 15 mL round bottom tubes (17x100 mm)
Expand
View Pricing
Z601152
Hettich
®
centrifuge adapters,
for 9-10 mL blood/urine tubes (15-16x92-102 mm) and 15 mL round bottom tubes (17x100 mm)
Expand
View Pricing
Z657883
Hettich
®
centrifuge adapters,
for ROTINA 38/38R and UNIVERSAL 320/320R, Adapter for 50 mL tube 34x100 mm
Expand
View Pricing
Z623598
Hettich
®
centrifuge adapters,
for ROTINA 38/38R, ROTINA 420/420R and ROTANTA 460/460R, for 4.5-5 mL blood/urine tubes (11x92 mm) and 7 mL round bottom tubes (12x100 mm)
Expand
View Pricing
Z641987
Hettich
®
centrifuge adapters,
for ROTINA 38/38R, ROTINA 420/420R and ROTANTA 460/460R Centrifuges, Adapter for 8.5-15 mL tubes (16-17x100 mm)
Expand
View Pricing
Z642002
Hettich
®
centrifuge adapters,
for ROTINA 38/38R, ROTINA 420/420R and ROTANTA 460/460R, Adapter for 25 mL tubes (24x100 mm)
Expand
View Pricing
Z601357
Hettich
®
centrifuge adapters,
adapter for 0.4 mL tubes 6x45 mm
Expand
View Pricing
Z601373
Hettich
®
centrifuge adapters,
adapter for 1.5-2.0 mL tubes 11x38 mm
Expand
View Pricing