Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hettich
®
adapter for 15 mL conical tubes
Synonym(s):
Hettich
®
centrifuge adapters
Products
Product Category
adapters (3)
Manufacturer
Package Size
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
Z728268
Hettich
®
adapter for 15 mL conical tubes,
for UNIVERSAL 320/320R and ROTOFIX 32A, Adapter for 4 x 15ml Conical tubes 17 x 120mm
Expand
View Pricing
Z728489
Hettich
®
adapter for 15 mL conical tubes,
for ROTANTA 460/460R Centrifuge, Adapter for 12 x 15 mL Conical Tubes 17 x 120mm
Expand
View Pricing
Z728802
Hettich
®
adapter for 15 mL conical tubes,
for ROTANTA 460/460R and ROTINA 38/38R Centrifuges, Adapter for 7 x 15 mL Conical Tubes 17 x 120 mm
Expand
View Pricing