Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Hastealloy
®
c-22
Products
Manufacturer
Physical Form
Length (mm)
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
GF29539705
Hastealloy
®
c-22,
rod, Ni59%/Cr22%/Mo13%/Fe 3%/W 3, 50.8 mm diameter, length 50 mm, temper annealed
Expand
View Pricing
GF94480487
Hastealloy
®
c-22,
rod, 6.35 mm diameter, Ni59%/Cr22%/Mo13%/Fe 3%/W 3%, length 100 mm, temper annealed
Expand
View Pricing
GF75372396
Hastealloy
®
c-22,
wire, 1.0 mm diameter, Ni59%/Cr22%/Mo13%/Fe 3%/W 3%, size 200 mm, temper annealed
Expand
View Pricing
GF03077906
Hastealloy
®
c-22,
rod, Ni59%/Cr22%/Mo13%/Fe 3%/W 3, 50.8 mm diameter, length 25 mm, temper annealed
Expand
View Pricing
GF18229978
Hastealloy
®
c-22,
rod, Ni59%/Cr22%/Mo13%/Fe 3%/W 3, 6.35 mm diameter, length 1000 mm, temper annealed
Expand
View Pricing
GF15959223
Hastealloy
®
c-22,
rod, Ni59%/Cr22%/Mo13%/Fe 3%/W 3, 6.35 mm diameter, length 200 mm, temper annealed
Expand
View Pricing
GF64480046
Hastealloy
®
c-22,
rod, 50.8 mm diameter, Ni59%/Cr22%/Mo13%/Fe 3%/W 3%, length 100 mm, temper annealed
Expand
View Pricing
GF40322393
Hastealloy
®
c-22,
wire, Ni59%/Cr22%/Mo13%/Fe 3%/W 3, 1.0 mm diameter, size 1110 mm, temper annealed
Expand
View Pricing
GF50151518
Hastealloy
®
c-22,
wire, Ni59%/Cr22%/Mo13%/Fe 3%/W 3, 1.0 mm diameter, size 500 mm, temper annealed
Expand
View Pricing
GF62176743
Hastealloy
®
c-22,
wire, 1.0 mm diameter, Ni59%/Cr22%/Mo13%/Fe 3%/W 3%, size 100 mm, temper annealed
Expand
View Pricing
GF81657748
Hastealloy
®
c-22,
rod, Ni59%/Cr22%/Mo13%/Fe 3%/W 3%, length 500 mm, 6.35 mm diameter, temper annealed
Expand
View Pricing