Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Harmine hydrochloride
Synonym(s):
7-Methoxy-1-methyl-9H-pyrido[3,4-b]indole hydrochloride
Empirical Formula (Hill Notation):
C
13
H
12
N
2
O · HCl
CAS Number:
343-27-1
Molecular Weight:
248.71
EC Number:
206-443-8
Products
Product Category
alkaloids (1)
inhibitors (1)
metabolites (1)
natural products (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Application
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
SMB00461
Harmine hydrochloride,
≥98% (HPLC)
Expand
View Pricing