Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Gly-Gly
Synonym(s):
Diglycine, Glycyl-glycine
Linear Formula:
NH
2
CH
2
CONHCH
2
COOH
CAS Number:
556-50-3
Molecular Weight:
132.12
EC Number:
209-127-8
Products
Product Category
buffers (4)
Brand
Sigma-Aldrich (4)
Color
Feature
Formula Weight
Gene Alias
Melting Point (°C)
pH Value
Physical Form
pK Value
Product Line
Purity
Quality Segment
Application
Special Grade
BioPerformance Certified (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
G1002
Gly-Gly,
≥99% (titration)
Expand
View Pricing
G3915
Gly-Gly,
BioPerformance Certified, suitable for cell culture, ≥99%
Expand
View Pricing
50199
Gly-Gly,
≥99.5% (NT), BioUltra
Expand
View Pricing
50200
Gly-Gly,
≥99.0% (NT), suitable for ligand binding assays, BioXtra
Expand
View Pricing