Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Glyceryl tridodecanoate
Synonym(s):
1,2,3-Tridodecanoylglycerol, 1,2,3-Trilauroylglycerol, Glycerol trilaurate, Glyceryl trilaurate, Tridodecanoin, Trilaurin
Linear Formula:
[CH
3
(CH
2
)
10
COOCH
2
]
2
CHOCO(CH
2
)
10
CH
3
CAS Number:
538-24-9
Molecular Weight:
639.00
EC Number:
208-687-0
Products
Product Category
glycerides (1)
lipids (1)
metabolites (1)
Brand
Sigma-Aldrich (1)
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
T4891
Glyceryl tridodecanoate,
≥99%
Expand
View Pricing