
Galactosamine hydrochloride

Synonym(s):
D-(+)-Galactosamine hydrochloride, 2-Amino-2-deoxy-D-galactopyranose hydrochloride, D-Chondrosamine hydrochloride, D-Galactosamine hydrochloride, Chondrosamine
Empirical Formula (Hill Notation):
C6H13NO5 · HCl
CAS Number:
Molecular Weight:
215.63
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2