Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Freezer box
Products
Product Category
boxes (3)
Manufacturer
Length (in)
Color
Package Size
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
Z744119
Freezer box,
W × L 2 in. × 2 in., to hold, 81 tube(s)
Expand
View Pricing
Z744120
Freezer box,
W × L 2 in. × 2 in., to hold, 100 tube(s)
Expand
View Pricing
Z744122
Freezer box,
W × L 3 in. × 3 in., without dividers
Expand
View Pricing
Z744121
Freezer box,
W × L 2 in. × 2 in., without dividers
Expand
View Pricing