
Fosinopril Related Compound A

Synonym(s):
Fosinoprilat, (4S)-4-Cyclohexyl-1-{[hydroxy(4-phenylbutyl)phosphinyl]acetyl}-L-proline
Empirical Formula (Hill Notation):
C23H34NO5P
CAS Number:
Molecular Weight:
435.49
Products
Brand
Formula Weight
Manufacturer
Format
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2