
Flonicamid

Synonym(s):
N-(Cyanomethyl)-4-(trifluoromethyl)nicotinamide, N-Cyanomethyl-4-trifluoromethyl-3-pyridinecarboxamide
Empirical Formula (Hill Notation):
C9H6F3N3O
CAS Number:
Molecular Weight:
229.16
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2