Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Fecralloy
®
- iron/chromium
Products
Manufacturer
Physical Form
Length (m)
Length (mm)
Filter Products
Showing 1-17 of 17
Compare
Product Number
Description
Pricing
GF08413781
Fecralloy
®
- iron/chromium,
rod, Fe 72.8%/Cr 22%/Al 5%/Y 0.1%/Zr 0.1%, 50.8 mm diameter, length 50 mm, condition hot rolled (black)
Expand
View Pricing
GF77027890
Fecralloy
®
- iron/chromium,
foil, thickness 0.05 mm, Fe72.8%/Cr22%/Al 5%/Y 0.1%/Zr 0.1%, size 100 × 100 mm, temper as rolled
Expand
View Pricing
GF83188660
Fecralloy
®
- iron/chromium,
foil, Fe72.8%/Cr22%/Al 5%/Y 0.1%/Zr 0.1%, thickness 0.05 mm, size 50 × 50 mm, temper as rolled
Expand
View Pricing
GF05673090
Fecralloy
®
- iron/chromium,
rod, Fe 72.8%/Cr 22%/Al 5%/Y 0.1%/Zr 0.1%, 36 mm diameter, length 200 mm, condition ground
Expand
View Pricing
GF22620710
Fecralloy
®
- iron/chromium,
rod, Fe 72.8%/Cr 22%/Al 5%/Y 0.1%/Zr 0.1%, 36 mm diameter, length 100 mm, condition ground
Expand
View Pricing
GF29144527
Fecralloy
®
- iron/chromium,
wire, Fe 72.8%/Cr 22%/Al 5%/Y 0.1%/Zr 0.1%, 1.6 mm diameter, length 50 m, temper annealed
Expand
View Pricing
GF34872116
Fecralloy
®
- iron/chromium,
wire, Fe 72.8%/Cr 22%/Al 5%/Y 0.1%/Zr 0.1%, 1.6 mm diameter, length 5 m, temper annealed
Expand
View Pricing
GF77949590
Fecralloy
®
- iron/chromium,
foil, thickness 0.05 mm, Fe72.8%/Cr22%/Al 5%/Y 0.1%/Zr 0.1%, size 250 × 250 mm, temper as rolled
Expand
View Pricing
GF73760070
Fecralloy
®
- iron/chromium,
wire, Fe72.8%/Cr22%/Al 5%/Y 0.1%/Zr 0.1%, 1.6 mm diameter, temper annealed, length 20 m
Expand
View Pricing
GF04540137
Fecralloy
®
- iron/chromium,
rod, Fe 72.8%/Cr 22%/Al 5%/Y 0.1%/Zr 0.1%, 75 mm diameter, length 50 mm, condition hot rolled (black)
Expand
View Pricing
GF67933907
Fecralloy
®
- iron/chromium,
rod, 75 mm diameter, Fe72.8%/Cr22%/Al 5%/Y 0.1%/Zr 0.1%, length 100 mm, condition hot rolled (black)
Expand
View Pricing
GF23515332
Fecralloy
®
- iron/chromium,
rod, Fe 72.8%/Cr 22%/Al 5%/Y 0.1%/Zr 0.1%, 36 mm diameter, length 50 mm, condition ground
Expand
View Pricing
GF71033341
Fecralloy
®
- iron/chromium,
wire, 1.6 mm diameter, Fe72.8%/Cr22%/Al 5%/Y 0.1%/Zr 0.1%, length 100 m, temper annealed
Expand
View Pricing
GF12913130
Fecralloy
®
- iron/chromium,
rod, Fe 72.8%/Cr 22%/Al 5%/Y 0.1%/Zr 0.1%, 67 mm diameter, length 100 mm, condition hot rolled (black)
Expand
View Pricing
GF92715813
Fecralloy
®
- iron/chromium,
rod, 50.8 mm diameter, Fe72.8%/Cr22%/Al 5%/Y 0.1%/Zr 0.1%, length 100 mm, condition hot rolled (black)
Expand
View Pricing
GF44001100
Fecralloy
®
- iron/chromium,
rod, Fe 72.8%/Cr 22%/Al 5%/Y 0.1%/Zr 0.1%, 67 mm diameter, length 50 mm, condition hot rolled (black)
Expand
View Pricing
GF84649630
Fecralloy
®
- iron/chromium,
wire, 1.6 mm diameter, Fe72.8%/Cr22%/Al 5%/Y 0.1%/Zr 0.1%, length 10 m, temper annealed
Expand
View Pricing