Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Exact-Temp Coated bottle thermometer with 2 point DAkkS-Calibration
Products
Color
Length (mm)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
Z742076
Exact-Temp Coated bottle thermometer with 2 point DAkkS-Calibration,
for incubator, temperature +15 to +30 °C, precision: ±0.1 °C
Expand
View Pricing
Z742072
Exact-Temp Coated bottle thermometer with 2 point DAkkS-Calibration,
for refrigerator, temperature -5 to +15 °C, precision: ±0.1 °C
Expand
View Pricing