
Ethylene glycol monomethyl ether

Synonym(s):
2-Methoxyethanol, Ethylene glycol monomethyl ether, Methyl glycol
Linear Formula:
CH3OCH2CH2OH
CAS Number:
Molecular Weight:
76.09
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing