Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ethyl azidoacetate solution
Linear Formula:
N
3
CH
2
COOCH
2
CH
3
CAS Number:
637-81-0
Molecular Weight:
129.12
Products
Product Category
building blocks (3)
Brand
Sigma-Aldrich (3)
Formula Weight
Physical Form
Markush Class
Markush Group
Reaction Type
Special Grade
Purum (3)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
77213
Ethyl azidoacetate solution,
~25% in toluene (NMR)
Expand
View Pricing
93528
Ethyl azidoacetate solution,
~25% in ethanol (NMR)
Expand
View Pricing
88539
Ethyl azidoacetate solution,
~30% in methylene chloride (NMR)
Expand
View Pricing