Skip to Content
Merck
Ethoxycarbonyl isothiocyanate 98%

Ethoxycarbonyl isothiocyanate

Ethoxycarbonyl isothiocyanate 98%
Synonym(s):
Ethyl isothiocyanatoformate
Linear Formula:
SCNCOOCH2CH3
CAS Number:
Molecular Weight:
131.15
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing