
Estropipate

Synonym(s):
3-(Sulfooxy)-estra-1,3,5(10)-trien-17-one : piperazine (1:1), Estrone sulfate piperazine salt, Harmogen, Ogen, Estropipate
Empirical Formula (Hill Notation):
C18H22O5S·C4H10N2
CAS Number:
Molecular Weight:
436.56
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-4 of 4