Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
EPPENDORF TWIN.TEC
®
PCR PLATES 384
Synonym(s):
Eppendorf twin.tec
®
PCR Plates 384
Products
Product Category
plates (3)
Manufacturer
Binding Type
Wells
Color
Package Size
Sterility
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
EP0030129547
EPPENDORF TWIN.TEC
®
PCR PLATES 384,
PCR Clean, colorless, pkg of 25 ×
Expand
View Pricing
EP0030129628
EPPENDORF TWIN.TEC
®
PCR PLATES 384,
skirted, Forensic DNA Grade, working volume 40 μL, colorless, pack of 10 (individually wrapped)
Expand
View Pricing
EP0030113578
EPPENDORF TWIN.TEC
®
PCR PLATES 384,
skirt, PCR clean, working volume 40 μL, colorless (with 2D SafeCode), pkg of 25 ea
Expand
View Pricing