Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
Research plus G, Basic
Products
Color
Manufacturer
Volume (ml)
Volume (µL)
Package Size
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
EP3124000016
Eppendorf
®
Research plus G, Basic,
single-channel, fixed, volume 10 μL, medium gray
Expand
View Pricing
EP3123000268
Eppendorf
®
Research plus G, Basic,
single-channel, variable, volume 100-1000 μL, blue
Expand
View Pricing
EP3123000276
Eppendorf
®
Research plus G, Basic,
single-channel, variable, volume 0.5-5 mL, purple
Expand
View Pricing
EP3123000250
Eppendorf
®
Research plus G, Basic,
single-channel, variable, volume 20-200 μL, yellow
Expand
View Pricing
EP3123000284
Eppendorf
®
Research plus G, Basic,
single-channel, variable, volume 1-10 mL, turquoise
Expand
View Pricing
EP3123000225
Eppendorf
®
Research plus G, Basic,
single-channel, variable, volume -10 μL, medium gray
Expand
View Pricing
EP3123000241
Eppendorf
®
Research plus G, Basic,
single-channel, variable, volume 10-100 μL, yellow
Expand
View Pricing
EP3123000233
Eppendorf
®
Research plus G, Basic,
single-channel, variable, volume 2-20 μL, yellow
Expand
View Pricing
EP3123000217
Eppendorf
®
Research plus G, Basic,
single-channel, variable, volume 0.1-2.5 μL, dark gray
Expand
View Pricing
EP3123000306
Eppendorf
®
Research plus G, Basic,
single-channel, variable, volume 30-300 μL, orange
Expand
View Pricing
EP3123000349
Eppendorf
®
Research plus G, Basic,
single-channel, variable, volume 0.25-2.5 mL, red, without tip box
Expand
View Pricing
EP3123000292
Eppendorf
®
Research plus G, Basic,
single-channel, variable, volume 2-20 μL, light gray
Expand
View Pricing