Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Eppendorf
®
epT.I.P.S.
®
G Reload 384
Products
Product Category
pipette tips (4)
Color
Manufacturer
Package Size
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
EP0030076052
Eppendorf
®
epT.I.P.S.
®
G Reload 384,
Eppendorf Quality
™
, tip capacity × L 0.5-100 μL × 52.8 mm, light yellow tip, case of 3,840 ea (10 trays x 384 tips)
Expand
View Pricing
EP0030076010
Eppendorf
®
epT.I.P.S.
®
G Reload 384,
PCR clean, tip capacity × L 0.5-100 μL × 53 mm, light yellow tip, case of 3,840 ea (10 trays x 384 tips)
Expand
View Pricing
EP0030076044
Eppendorf
®
epT.I.P.S.
®
G Reload 384,
Eppendorf Quality
™
, tip capacity × L 0.1-20 μL × 42 mm, white tip, case of 3,840 ea (10 trays x 384 tips)
Expand
View Pricing
EP0030076001
Eppendorf
®
epT.I.P.S.
®
G Reload 384,
PCR clean, tip capacity × L 0.1-20 μL × 42 mm, white tip, case of 3,840 ea (10 trays x 384 tips)
Expand
View Pricing